The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019371.1 | NP_062244.1 | 4 - 5 | 1224 | 99 |
GenBank mRNA | U06713.1 | - | 4 - 5 | 1224 | 99 |