Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Taf7l
Entrez Gene ID
363493
Gene Name
TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
RGD1564898
Chromosome Location
Chr.X: 105292338 - 105308554 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135877.1 NP_001129349.1 11 - 12 1351 127

View Full Product Details