Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dmrtc1c1
Entrez Gene ID
363483
Gene Name
DMRT-like family C1c1
Gene Aliases
Dmrtc1c, Dmrtc1c2
Chromosome Location
Chr.X: 72684315 - 72691535 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014222.2 NP_001014244.2 4 - 5 625 70
GenBank mRNA BC079417.1 - 4 - 5 611 70
CV112810.1 - 4 - 5 625 70

View Full Product Details