Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cstf2
Entrez Gene ID
683927
Gene Name
cleavage stimulation factor, 3' pre-RNA subunit 2
Gene Aliases
-
Chromosome Location
Chr.X: 104882727 - 104909746 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131014.1 NP_001124486.1 13 - 14 1739 101
XM_008773440.1 XP_008771662.1 15 - 16 1991 101
GenBank mRNA BC168251.1 - 13 - 14 1739 101

View Full Product Details