Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan6
Entrez Gene ID
302313
Gene Name
tetraspanin 6
Gene Aliases
-
Chromosome Location
Chr.X: 104719930 - 104729563 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100672.1 NP_001094142.1 1 - 2 105 94
GenBank mRNA BC158651.1 - 1 - 2 105 94
FQ213424.1 - 1 - 2 162 94
FQ216491.1 - 1 - 2 128 94

View Full Product Details