Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tspan6
Entrez Gene ID
302313
Gene Name
tetraspanin 6
Gene Aliases
-
Chromosome Location
Chr.X: 104719930 - 104729563 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100672.1 NP_001094142.1 3 - 4 360 144
XM_008773408.1 XP_008771630.1 4 - 5 620 144
GenBank mRNA BC086430.1 - 2 - 3 200 144
BC158651.1 - 3 - 4 360 144
FQ213424.1 - 3 - 4 417 144
FQ216491.1 - 3 - 4 383 144

View Full Product Details