The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025645.1 | NP_001020816.1 | 2 - 3 | 401 | 84 |
XM_006230747.2 | XP_006230809.1 | 2 - 3 | 625 | 84 | |
XM_008760106.1 | XP_008758328.1 | 2 - 3 | 625 | 84 | |
XM_008760107.1 | XP_008758329.1 | 2 - 3 | 625 | 84 | |
GenBank mRNA | BC098853.1 | - | 2 - 3 | 401 | 84 |