Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Poglut1
Entrez Gene ID
288091
Gene Name
protein O-glucosyltransferase 1
Gene Aliases
Ktelc1, RGD1306248
Chromosome Location
Chr.11: 64761493 - 64788210 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100652.1 NP_001094122.1 7 - 8 830 142
GenBank mRNA BC091408.1 - 6 - 7 602 142

View Full Product Details