The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013865.1 | NP_001013887.1 | 8 - 9 | 1576 | 64 |
XM_008768717.1 | XP_008766939.1 | 7 - 8 | 1531 | 64 | |
GenBank mRNA | BC085845.1 | - | 8 - 9 | 1576 | 64 |