Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fstl1
Entrez Gene ID
79210
Gene Name
follistatin-like 1
Gene Aliases
Frp, Fstl
Chromosome Location
Chr.11: 65792192 - 65845375 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024369.2 NP_077345.1 10 - 11 962 69
GenBank mRNA BC087014.1 - 10 - 11 962 69
U06864.1 - 10 - 11 937 69

View Full Product Details