The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107090.2 | NP_001100560.1 | 12 - 13 | 2641 | 91 |
XM_006248450.2 | XP_006248512.1 | 11 - 12 | 2696 | 91 | |
XM_006248451.2 | XP_006248513.1 | 12 - 13 | 2272 | 91 | |
XM_006248452.2 | XP_006248514.1 | 12 - 13 | 2153 | 91 | |
XM_008768726.1 | XP_008766948.1 | 12 - 13 | 2422 | 91 | |
GenBank mRNA | BC168952.1 | - | 12 - 13 | 2641 | 91 |