Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Muc4
Entrez Gene ID
303887
Gene Name
mucin 4, cell surface associated
Gene Aliases
ASGP-1, Psmc
Chromosome Location
Chr.11: 71242973 - 71285227 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248471.2 XP_006248533.1 13 - 14 5050 88
XM_008768775.1 XP_008766997.1 13 - 14 4690 88
XM_008768776.1 XP_008766998.1 13 - 14 3958 88
XM_008768777.1 XP_008766999.1 13 - 14 3227 88
GenBank mRNA U06752.1 - 14 - 15 3428 88

View Full Product Details