Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap17b
Entrez Gene ID
313434
Gene Name
A kinase (PRKA) anchor protein 17B
Gene Aliases
RGD1566363
Chromosome Location
Chr.X: 123217410 - 123246049 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002730183.4 XP_002730229.1 1 - 2 766 99

View Full Product Details