Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl13
Entrez Gene ID
313445
Gene Name
kelch-like family member 13
Gene Aliases
-
Chromosome Location
Chr.X: 121578965 - 121735007 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257445.2 XP_006257507.1 3 - 4 331 84

View Full Product Details