Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ldha
Entrez Gene ID
24533
Gene Name
lactate dehydrogenase A
Gene Aliases
Ldh1
Chromosome Location
Chr.1: 102900288 - 102909713 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017025.1 NP_058721.1 2 - 3 231 150
XM_006229232.2 XP_006229294.1 2 - 3 399 150
GenBank mRNA BC084698.1 - 1 - 2 133 150
BC168737.1 - 2 - 3 230 150
FQ214854.1 - 2 - 3 231 150
X01964.1 - 2 - 3 231 150

View Full Product Details