Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcna7
Entrez Gene ID
365241
Gene Name
potassium channel, voltage gated shaker related subfamily A, member 7
Gene Aliases
-
Chromosome Location
Chr.1: 101397828 - 101403320 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108914.1 NP_001102384.1 1 - 2 1072 134

View Full Product Details