Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fxyd7
Entrez Gene ID
63848
Gene Name
FXYD domain-containing ion transport regulator 7
Gene Aliases
-
Chromosome Location
Chr.1: 89474601 - 89484068 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022008.1 NP_071291.1 4 - 5 268 77
GenBank mRNA AW914994.1 - 4 - 5 268 77

View Full Product Details