Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc7a10
Entrez Gene ID
114518
Gene Name
solute carrier family 7 (neutral amino acid transporter light chain, asc system), member 10
Gene Aliases
Asc-1
Chromosome Location
Chr.1: 91432952 - 91449504 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053726.2 NP_446178.2 8 - 9 1186 72
GenBank mRNA AB126813.1 - 8 - 9 1186 72
BC127467.1 - 8 - 9 1157 72

View Full Product Details