Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rtn2
Entrez Gene ID
308410
Gene Name
reticulon 2
Gene Aliases
RTN2-B
Chromosome Location
Chr.1: 80195594 - 80208449 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201562.3 NP_963856.1 3 - 4 545 62
GenBank mRNA AY279211.1 - 3 - 4 545 62
BC129079.1 - 3 - 4 660 62

View Full Product Details