Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Krtdap
Entrez Gene ID
65186
Gene Name
keratinocyte differentiation associated protein
Gene Aliases
Kdap
Chromosome Location
Chr.1: 89232442 - 89234198 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB011028.1 - - 218 111

View Full Product Details