Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Josd2
Entrez Gene ID
292876
Gene Name
Josephin domain containing 2
Gene Aliases
RGD1307305
Chromosome Location
Chr.1: 100473419 - 100477352 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106256.2 NP_001099726.2 1 - 2 127 58
GenBank mRNA DV214517.1 - 1 - 2 127 58

View Full Product Details