The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106256.2 | NP_001099726.2 | 4 - 5 | 604 | 85 |
XM_006228996.2 | XP_006229058.1 | 5 - 6 | 886 | 85 | |
XM_006228997.2 | XP_006229059.1 | 5 - 6 | 883 | 85 | |
XM_006228998.2 | XP_006229060.1 | 4 - 5 | 825 | 85 | |
GenBank mRNA | CK842299.1 | - | 1 - 2 | 85 |