Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aspdh
Entrez Gene ID
292875
Gene Name
aspartate dehydrogenase domain containing
Gene Aliases
RGD1310111
Chromosome Location
Chr.1: 100471038 - 100473327 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009643.2 NP_001009643.2 1 - 2 84 84
GenBank mRNA BC088252.1 - 1 - 2 84 84

View Full Product Details