Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Npas1
Entrez Gene ID
308387
Gene Name
neuronal PAS domain protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 78420646 - 78441841 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107479.1 NP_001100949.1 9 - 10 1278 113
XM_006228413.2 XP_006228475.1 10 - 11 1406 113

View Full Product Details