Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klk11
Entrez Gene ID
292849
Gene Name
kallikrein related-peptidase 11
Gene Aliases
-
Chromosome Location
Chr.1: 99712108 - 99716648 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106252.1 NP_001099722.1 5 - 6 813 118

View Full Product Details