The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107496.1 | NP_001100966.1 | 2 - 3 | 323 | 68 |
XM_006228621.2 | XP_006228683.1 | 2 - 3 | 487 | 68 | |
XM_006228622.2 | XP_006228684.1 | 2 - 3 | 487 | 68 | |
XM_006228623.2 | XP_006228685.1 | 2 - 3 | 487 | 68 | |
XM_006228624.2 | XP_006228686.1 | 2 - 3 | 338 | 68 |