Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fosb
Entrez Gene ID
100360880
Gene Name
FBJ osteosarcoma oncogene B
Gene Aliases
fra-2
Chromosome Location
Chr.1: 80214472 - 80221931 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256509.1 NP_001243438.1 2 - 3 766 89

View Full Product Details