Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2f8
Entrez Gene ID
308607
Gene Name
E2F transcription factor 8
Gene Aliases
E2F-8, RGD1308091
Chromosome Location
Chr.1: 104185436 - 104203684 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006229263.2 XP_006229325.1 6 - 7 1772 148
GenBank mRNA BC099080.1 - 1 - 2 121 148

View Full Product Details