Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lin37
Entrez Gene ID
292787
Gene Name
lin-37 DREAM MuvB core complex component
Gene Aliases
RGD1308901
Chromosome Location
Chr.1: 89010257 - 89014484 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106245.1 NP_001099715.1 3 - 4 327 66
XM_006228769.2 XP_006228831.1 3 - 4 748 66
GenBank mRNA FQ233946.1 - 3 - 4 295 66

View Full Product Details