The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025014.1 | NP_001020185.1 | 4 - 5 | 439 | 77 |
XM_006248066.2 | XP_006248128.1 | 5 - 6 | 607 | 77 | |
XM_006248067.2 | XP_006248129.1 | 5 - 6 | 693 | 77 | |
XM_006248068.2 | XP_006248130.1 | 4 - 5 | 559 | 77 | |
XM_006248069.2 | XP_006248131.1 | 4 - 5 | 445 | 77 | |
GenBank mRNA | BC091349.1 | - | 4 - 5 | 439 | 77 |