The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024400.2 | NP_077376.2 | 8 - 9 | 2589 | 80 |
GenBank mRNA | AF149118.1 | - | 8 - 9 | 2602 | 80 |
AF304446.1 | - | 8 - 9 | 2160 | 80 | |
BC080237.1 | - | 8 - 9 | 2589 | 80 |