Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lipi
Entrez Gene ID
288322
Gene Name
lipase, member I
Gene Aliases
Liph
Chromosome Location
Chr.11: 13960246 - 13999869 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105899.1 NP_001099369.1 2 - 3 205 128

View Full Product Details