Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Whamm
Entrez Gene ID
293057
Gene Name
WAS protein homolog associated with actin, golgi membranes and microtubules
Gene Aliases
RGD1563506, Whdc1
Chromosome Location
Chr.1: 143401986 - 143430356 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130728.1 NP_001124200.1 2 - 3 806 86

View Full Product Details