The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025681.1 | NP_001020852.2 | 3 - 4 | 454 | 91 |
XM_008759515.1 | XP_008757737.1 | 3 - 4 | 1346 | 91 | |
XM_008759516.1 | XP_008757738.1 | 3 - 4 | 873 | 91 | |
XM_008759517.1 | XP_008757739.1 | 3 - 4 | 1346 | 91 | |
XM_008759518.1 | XP_008757740.1 | 3 - 4 | 1343 | 91 | |
GenBank mRNA | BC097953.1 | - | 3 - 4 | 454 | 91 |