Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc23
Entrez Gene ID
308708
Gene Name
tetratricopeptide repeat domain 23
Gene Aliases
RGD1310409
Chromosome Location
Chr.1: 128606340 - 128687898 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025681.1 NP_001020852.2 8 - 9 1143 99
XM_008759515.1 XP_008757737.1 8 - 9 2035 99
XM_008759516.1 XP_008757738.1 8 - 9 1562 99
GenBank mRNA BC097953.1 - 8 - 9 1143 99

View Full Product Details