Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snrpa1
Entrez Gene ID
100361269
Gene Name
small nuclear ribonucleoprotein polypeptide A'
Gene Aliases
-
Chromosome Location
Chr.1: 126961389 - 126974453 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271323.1 NP_001258252.1 2 - 3 301 102
XM_006229323.2 XP_006229385.1 2 - 3 460 102
GenBank mRNA FM044635.1 - 2 - 3 285 102
FQ219673.1 - 2 - 3 285 102
FQ231357.1 - 2 - 3 301 102
FQ232647.1 - 2 - 3 286 102

View Full Product Details