The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108915.1 | NP_001102385.1 | 5 - 6 | 1529 | 95 |
XM_006229349.2 | XP_006229411.1 | 6 - 7 | 1731 | 95 | |
XM_006229350.2 | XP_006229412.1 | 6 - 7 | 1697 | 95 | |
XM_008759545.1 | XP_008757767.1 | 5 - 6 | 2675 | 95 |