Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fah
Entrez Gene ID
29383
Gene Name
fumarylacetoacetate hydrolase
Gene Aliases
-
Chromosome Location
Chr.1: 146713663 - 146736339 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017181.2 NP_058877.1 11 - 12 1060 103
GenBank mRNA BC076381.1 - 11 - 12 1060 103
FQ219602.1 - 11 - 12 1019 103
M77694.1 - 11 - 12 982 103

View Full Product Details