Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh3gl3
Entrez Gene ID
81921
Gene Name
SH3-domain GRB2-like 3
Gene Aliases
SH3P13, Sh3d2c1
Chromosome Location
Chr.1: 144069641 - 144200402 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031238.2 NP_112517.1 4 - 5 680 59
XM_006229480.2 XP_006229542.1 4 - 5 468 59
XM_006229481.2 XP_006229543.1 4 - 5 561 59
XM_006229483.2 XP_006229545.1 4 - 5 667 59
XM_006229485.1 XP_006229547.1 2 - 3 211 59
GenBank mRNA AB008159.2 - 1 - 2 125 59
AF227439.1 - 4 - 5 683 59
BC078800.1 - 4 - 5 683 59

View Full Product Details