The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001115039.1 | NP_001108511.1 | 4 - 5 | 491 | 83 |
GenBank mRNA | BC158881.1 | - | 4 - 5 | 491 | 83 |