Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nkx6-3
Entrez Gene ID
685102
Gene Name
NK6 homeobox 3
Gene Aliases
-
Chromosome Location
Chr.16: 73665269 - 73670482 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109455.1 NP_001102925.1 1 - 2 697 73

View Full Product Details