Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lsm1
Entrez Gene ID
364624
Gene Name
LSM1, U6 small nuclear RNA associated
Gene Aliases
-
Chromosome Location
Chr.16: 71046475 - 71057883 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108876.1 NP_001102346.1 1 - 2 397 113
GenBank mRNA FQ216033.1 - 1 - 2 167 113
FQ220497.1 - 1 - 2 265 113

View Full Product Details