Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Erich1
Entrez Gene ID
306622
Gene Name
glutamate-rich 1
Gene Aliases
RGD1559872
Chromosome Location
Chr.16: 80601888 - 80667773 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127680.1 NP_001121152.1 5 - 6 984 102
XM_006253395.1 XP_006253457.1 5 - 6 1060 102
GenBank mRNA BC167071.1 - 5 - 6 980 102
FQ227386.1 - 5 - 6 979 102

View Full Product Details