The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024773.1 | NP_001019944.1 | 3 - 4 | 605 | 79 |
GenBank mRNA | BC097435.1 | - | 3 - 4 | 605 | 79 |