Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Psmd13
Entrez Gene ID
365388
Gene Name
proteasome (prosome, macropain) 26S subunit, non-ATPase, 13
Gene Aliases
-
Chromosome Location
Chr.1: 213636159 - 213648499 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108925.2 NP_001102395.1 10 - 11 856 56
GenBank mRNA BC158732.1 - 10 - 11 856 56

View Full Product Details