Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Naa40
Entrez Gene ID
361718
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Gene Aliases
Nat11, RGD1565838
Chromosome Location
Chr.1: 222478021 - 222495382 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108518.1 NP_001101988.1 7 - 8 807 83

View Full Product Details