Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dak
Entrez Gene ID
361730
Gene Name
dihydroxyacetone kinase 2 homolog (S. cerevisiae)
Gene Aliases
RGD1311026
Chromosome Location
Chr.1: 226642901 - 226657698 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039031.1 NP_001034120.1 3 - 4 475 63
XM_006231056.2 XP_006231118.1 3 - 4 361 63
XM_006231057.2 XP_006231119.2 3 - 4 644 63
GenBank mRNA BC098925.1 - 3 - 4 475 63

View Full Product Details