Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pstk
Entrez Gene ID
361661
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
RGD1564300
Chromosome Location
Chr.1: 201950148 - 201960563 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271229.1 NP_001258158.1 5 - 6 917 110
GenBank mRNA BC168162.1 - 4 - 5 1151 110

View Full Product Details