Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpp30
Entrez Gene ID
685332
Gene Name
ribonuclease P/MRP 30 subunit
Gene Aliases
-
Chromosome Location
Chr.1: 254697247 - 254720093 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191083.1 NP_001178012.1 1 - 2 93 139

View Full Product Details