The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107565.1 | NP_001101035.1 | 3 - 4 | 603 | 71 |
XM_006230782.1 | XP_006230844.1 | 4 - 5 | 410 | 71 |